Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001788

Bilberry dry extract, HRS

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bilberry extract

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

matrix material

storage temp.

−20°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bilberry dry extract, HRS has been used as an EP reference standard:
  • For the identification of powdered herbal drug of dried bilberry fruit using thin layer chromatography technique (TLC).
  • For the determination of total anthocyanidins in fresh bilberry fruit dry extract using chromatography techniques.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bilberry fruit, dried
European Pharmacopoeia 10.3
European Pharmacopoeia II, 10(3) null
Fresh bilberry fruit dry extract, refined and standardised
European Pharmacopoeia 10.3
European Pharmacopoeia II, 10(3) null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service