Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0001473

Goserelin for NMR identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Goserelin acetate, [D-Ser(tBu)6,Azagly10]-LHRH acetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H84N18O14
CAS Number:
Molecular Weight:
1269.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

goserelin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)C[C@H](NC(=O)[C@@H](COC(C)(C)C)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](CO)NC(=O)[C@H](Cc2c[nH]c3ccccc23)NC(=O)[C@H](Cc4c[nH]cn4)NC(=O)[C@@H]5CCC(=O)N5)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N6CCC[C@H]6C(=O)NNC(N)=O

InChI

1S/C59H84N18O14.C2H4O2/c1-31(2)22-40(49(82)68-39(12-8-20-64-57(60)61)56(89)77-21-9-13-46(77)55(88)75-76-58(62)90)69-54(87)45(29-91-59(3,4)5)74-50(83)41(23-32-14-16-35(79)17-15-32)70-53(86)44(28-78)73-51(84)42(24-33-26-65-37-11-7-6-10-36(33)37)71-52(85)43(25-34-27-63-30-66-34)72-48(81)38-18-19-47(80)67-38;1-2(3)4/h6-7,10-11,14-17,26-27,30-31,38-46,65,78-79H,8-9,12-13,18-25,28-29H2,1-5H3,(H,63,66)(H,67,80)(H,68,82)(H,69,87)(H,70,86)(H,71,85)(H,72,81)(H,73,84)(H,74,83)(H,75,88)(H4,60,61,64)(H3,62,76,90);1H3,(H,3,4)/t38-,39-,40-,41-,42-,43-,44-,45+,46-;/m0./s1

InChI key

IKDXDQDKCZPQSZ-JHYYTBFNSA-N

Gene Information

human ... GNRHR(2798)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Ser(tBu)-Leu-Arg-Pro-azaGly-NH2

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Goserelin for NMR identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Goserelin is a synthetic analog of LHRH (leutinizing hormone releasing hormone). Goserelin is a LHRH agonist It is used in prostate cancer and breast cancer therapies and also in the treatment of endometriosis.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven P Schwendeman et al.
Journal of controlled release : official journal of the Controlled Release Society, 190, 240-253 (2014-06-15)
Biodegradable, injectable depot formulations for long-term controlled drug release have improved therapy for a number of drug molecules and led to over a dozen highly successful pharmaceutical products. Until now, success has been limited to several small molecules and peptides
Halle C F Moore et al.
The New England journal of medicine, 372(10), 923-932 (2015-03-05)
Ovarian failure is a common toxic effect of chemotherapy. Studies of the use of gonadotropin-releasing hormone (GnRH) agonists to protect ovarian function have shown mixed results and lack data on pregnancy outcomes. We randomly assigned 257 premenopausal women with operable
Jie Wang et al.
Molecular pharmaceutics, 12(3), 966-973 (2015-01-24)
A major barrier to successful oral delivery of peptide and protein molecules is their inherent instability in the lumen of the gastrointestinal tract. The aim of this study was to determine the stability of 17 disparate peptide drugs (insulin, calcitonin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service