Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001400

Vigabatrin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Vigabatrin, (R,S)-4-Amino-5-hexenoic acid, (±)-γ-Vinyl-GABA, (±)-4-Aminohexenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

vigabatrin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(CCC(O)=O)C=C

InChI

1S/C6H11NO2/c1-2-5(7)3-4-6(8)9/h2,5H,1,3-4,7H2,(H,8,9)

InChI key

PJDFLNIOAUIZSL-UHFFFAOYSA-N

Gene Information

human ... ABAT(18)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Vigabatrin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Irreversible GABA transaminase inhibitor. Increases intracellular concentration of GABA in nerve endings; possesses antiepileptic activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Carmant
Acta neurologica Scandinavica. Supplementum, (192)(192), 36-47 (2011-11-09)
Infantile spasms (IS) are a unique and severe form of epilepsy associated with poor neurologic and developmental outcomes. The refractory spasms and abnormal electroencephalogram (EEG) patterns associated with the condition are believed to have a progressively detrimental impact. Therefore, rapid
Melissa J Maguire et al.
Epilepsia, 51(12), 2423-2431 (2010-11-13)
Vigabatrin is an efficacious antiepileptic drug licensed as add-on therapy in refractory epilepsy and used in infantile spasms. Eight years after licensing, there emerged a strong and possibly causative association with bilateral visual field loss. We report a systematic review
E Ben-Menachem
Acta neurologica Scandinavica. Supplementum, (192)(192), 5-15 (2011-11-09)
Discovered more than three decades ago, vigabatrin is approved in more than 50 countries as adjunctive therapy for adult patients with refractory complex partial seizures who have responded inadequately to several alternative treatments and as monotherapy for pediatric patients aged
J M Pellock
Acta neurologica Scandinavica. Supplementum, (192)(192), 83-91 (2011-11-09)
Vigabatrin is an effective and well-tolerated antiepileptic drug (AED) for the treatment of refractory complex partial seizures (rCPS) and infantile spasms (IS), but its benefits must be evaluated in conjunction with its risk of retinopathy with the development of peripheral
Karla Hemming et al.
The Cochrane database of systematic reviews, 1(1), CD007302-CD007302 (2013-02-27)
Epilepsy is a common neurological condition which affects between 0.5% and 1% of the population. Approximately 30% of people with epilepsy do not respond to treatment with currently available drugs. The majority of these people have partial epilepsy. Vigabatrin is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service