Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001387

Candesartan cilexetil for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Candesartan cilexetil, 2-ethoxy-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-Benzimidazole-7-carboxylic acid 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, TCV 116, TCY 116

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34N6O6
CAS Number:
Molecular Weight:
610.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

candesartan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCOc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)-c5ccccc5-c6nnn[nH]6

InChI

1S/C33H34N6O6/c1-3-42-32-34-28-15-9-14-27(31(40)43-21(2)44-33(41)45-24-10-5-4-6-11-24)29(28)39(32)20-22-16-18-23(19-17-22)25-12-7-8-13-26(25)30-35-37-38-36-30/h7-9,12-19,21,24H,3-6,10-11,20H2,1-2H3,(H,35,36,37,38)

InChI key

GHOSNRCGJFBJIB-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Candesartan cilexetil for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Candesartan cilexetil is a non-peptide angiotensin (AT2) receptor antagonist.
Candesartan cilexetil is the prodrug form of the potent angiotensin II receptor antagonist, candesartan. The prodrug is cleaved by esterases within the intestine to liberate the active molecule.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2 Oral

Target Organs

Kidney,Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Greg L Plosker et al.
PharmacoEconomics, 24(12), 1249-1272 (2006-11-30)
The addition of candesartan cilexetil (Atacand, Amias, Blopress, Kenzen, Ratacand) to standard therapy for chronic heart failure (CHF) provided important clinical benefits at little or no additional cost in France, Germany and the UK, according to a detailed economic analysis
Peter A Meredith
Current medical research and opinion, 23(7), 1693-1705 (2007-06-26)
Therapeutic interventions that block the renin-angiotensin-aldosterone system (RAAS) have an important role in slowing the progression of cardiovascular risk actors to established cardiovascular diseases. In recent years, angiotensin receptor blockers (ARBs) have emerged as effective and well-tolerated alternatives to an
Naoki Ichikawa et al.
Neurological research, 37(2), 147-152 (2014-08-05)
The purpose of this study was to examine whether oral administration of an angiotensin II type 1 receptor blocker (ARB) inhibited in-stent neointimal hyperplasia in carotid arteries of hypercholesterolemic rabbits. Eleven male New Zealand white rabbits were subjected to endothelial
Neetu Singh et al.
International journal of stroke : official journal of the International Stroke Society, 9(5), 560-568 (2012-09-28)
Cerebral ischaemia results in enhanced expression of type 1 angiotensin receptor and oxidative stress. Free radicals due to oxidative stress lead to excessive DNA damage causing overactivation of poly (ADP-ribose) polymerase-1 resulting in neuronal death. Activation of both type 1
Alexander Joost et al.
Expert opinion on pharmacotherapy, 12(11), 1769-1780 (2011-06-10)
Candesartan cilexetil is one of the most often-used first-line drugs regarding the management of arterial hypertension. Moreover, this drug has proven its effectiveness in chronic heart failure and exerts beneficial effects in diabetes, stroke, dementia and atrial fibrillation. This review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service