Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0001322

Hydroxyethyl starch (high MW)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hydroxyethyl starch

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

starch

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O.OCCOC[C@H]2O[C@H](OCCO)[C@H](OCCO)[C@@H](OCCO)[C@@H]2OCCO

InChI

1S/C16H32O11.C6H12O6/c17-1-6-22-11-12-13(23-7-2-18)14(24-8-3-19)15(25-9-4-20)16(27-12)26-10-5-21;7-1-2-3(8)4(9)5(10)6(11)12-2/h12-21H,1-11H2;2-11H,1H2/t12-,13-,14+,15-,16+;2-,3-,4+,5-,6+/m11/s1

InChI key

DNZMDASEFMLYBU-RNBXVSKKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. For further information and support, including product information leaflets, please go to the website of the issuing Pharmacopoeia.

Application

Hydroxyethyl starch (high MW) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service