Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001318

Cefpodoxime proxetil for impurity H identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cefpodoxime proxetil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N5O9S2
CAS Number:
Molecular Weight:
557.60
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cefpodoxime

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[s]1c(nc(c1)\C(=N\OC)\C(=O)N[C@H]2[C@H]3SCC(=C(N3C2=O)C(=O)OC(OC(=O)OC(C)C)C)COC)N

InChI

1S/C21H27N5O9S2/c1-9(2)33-21(30)35-10(3)34-19(29)15-11(6-31-4)7-36-18-14(17(28)26(15)18)24-16(27)13(25-32-5)12-8-37-20(22)23-12/h8-10,14,18H,6-7H2,1-5H3,(H2,22,23)(H,24,27)/b25-13-/t10?,14-,18-/m1/s1

InChI key

LTINZAODLRIQIX-FBXRGJNPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cefpodoxime proxetil for impurity H identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amrita Bajaj et al.
Drug development and industrial pharmacy, 39(5), 635-645 (2012-05-09)
Lipid based drug delivery systems have gained prominence in last decade for drugs with dissolution rate limited oral bioavailability. To improve the solubility, permeability and oral bioavailability of cefpodoxime proxetil, β-lactam antibiotic. It is BCS Class IV drug having solubility
Md Salim Shakur et al.
Indian pediatrics, 44(11), 838-841 (2007-12-07)
In order to evaluate clinical and bacteriological efficacy of Cefpodoxime Proxetil (CP) in typhoid fever in comparison to cefixime (CF), we assessed 140 children with suspected typhoid fever. Fulfilling inclusion criteria finally 40 culture confirmed typhoid fever were allocated in
Hakki Yilmaz et al.
Current drug safety, 8(2), 145-147 (2013-07-13)
We report a case of acute interstitial nephritis (AIN) and immune hemolytic anemia (IHA) associated with cefpodoxime therapy. A patient with a recent history of cefpodoxime proxetil treatment presented with elevated serum creatinine, oliguria, nausea, vomiting, and dyspnea. Evidence of
Santosh V Gandhi et al.
Hindustan antibiotics bulletin, 51(1-4), 24-28 (2009-01-01)
Two accurate, precise, sensitive and economical procedures for simultaneous estimation of Cefpodoxime proxetil and Potassium clavulanate in tablet dosage form have been developed. The methods employed were absorbance correction method (I) and first order derivative spectroscopic method (II). The first
Abhijit A Date et al.
International journal of pharmaceutics, 329(1-2), 166-172 (2006-10-03)
Self-nanoemulsifying drug delivery systems (SNEDDS) were developed with the objective to overcome problems associated with the delivery of cefpodoxime proxetil (CFP), a poorly bioavailable high dose antibiotic having pH dependant solubility. Solubility of CFP in oily phases and surfactants was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service