Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001103

Dimenhydrinate for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dimenhydrinate, N-(2-Diphenylmethoxyethyl)-N,N-dimethylammonium 8-chlorotheophyllinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO · C7H7ClN4O2
CAS Number:
Molecular Weight:
469.96
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

dimenhydrinate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1C(=O)N(C)c2nc(Cl)[nH]c2C1=O.CN(C)CCOC(c3ccccc3)c4ccccc4

InChI

1S/C17H21NO.C7H7ClN4O2/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;1-11-4-3(9-6(8)10-4)5(13)12(2)7(11)14/h3-12,17H,13-14H2,1-2H3;1-2H3,(H,9,10)

InChI key

NFLLKCVHYJRNRH-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dimenhydrinate for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min-Beom Kim et al.
The Laryngoscope, 124(10), 2400-2403 (2014-05-02)
To investigate the characteristics of residual symptoms and to evaluate the effects of adjuvant vestibular suppressants on residual symptoms after successful canalith repositioning procedures (CRPs). Individual randomized controlled trial. One hundred fifty patients with idiopathic benign paroxysmal positional vertigo who
Alison G Halpert et al.
Neuroscience and biobehavioral reviews, 26(1), 61-67 (2002-02-12)
The over-the-counter anti-emetic dimenhydrinate (DMH) (Gravol or Dramamine) has been reported to be abused for non-medicinal purposes. Street drug users abuse DMH for the acute effects of euphoric sensations and hallucinations, while psychiatric patients abuse DMH for its anxiolytic or

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service