Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0001004

Dydrogesterone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

9β,10α-Pregna-4,6-diene-3,20-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

dydrogesterone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1CC[C@]2([C@H]3[C@H]([C@H]4[C@@]([C@H](CC4)C(=O)C)(CC3)C)C=CC2=C1)C

InChI

1S/C21H28O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4-5,12,16-19H,6-11H2,1-3H3/t16-,17+,18-,19+,20+,21+/m0/s1

InChI key

JGMOKGBVKVMRFX-HQZYFCCVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dydrogesterone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R H Foster et al.
Drugs & aging, 11(4), 309-332 (1997-10-29)
The focus of this review is hormone replacement therapy (HRT) with continuous oral 17 beta-estradiol (herein referred to as estradiol) 2 mg/day plus sequential oral dydrogesterone 10 or 20 mg/day for 14 days of each 28-day cycle. According to data
Alfred O Mueck et al.
Maturitas, 65 Suppl 1, S51-S60 (2009-10-20)
According to the Women's Health Initiative (WHI) study, the most important risks during combined hormone replacement therapy (HRT) are breast cancer, stroke and venous thromboembolism. To date, combinations of estradiol with natural progesterone or its retroisomer dydrogesterone are the only
Annette Queisser-Luft
Early human development, 85(6), 375-377 (2009-02-06)
Between 1977 and 2005, 28 cases of potential links between maternal dydrogesterone use during pregnancy and congenital birth defects were reported. The types of defects were very diverse, with no evidence of a pattern of abnormalities. The data do not
Christian J Gruber et al.
The Journal of steroid biochemistry and molecular biology, 97(5), 426-430 (2005-09-29)
The published evidence regarding the administration of dydrogesterone in the treatment of habitual abortion is summarised in this review. Habitual abortion is defined as the loss of three or more consecutive pregnancies without known maternal or foetal pathology. The immunology
Marta Borowiecka et al.
Journal of women's health (2002), 19(12), 2267-2270 (2010-11-09)
Oral hormone therapy (HT) and menopausal age are both prothrombotic risk factors. The aim of our study was to compare the hemostatic parameters in plasma of postmenopausal women after 6 months of oral HT with parameters of control (without treatment)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service