Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000858

Isoflurane

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H2ClF5O
CAS Number:
Molecular Weight:
184.49
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

isoflurane

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)C(Cl)OC(F)F

InChI

1S/C3H2ClF5O/c4-1(3(7,8)9)10-2(5)6/h1-2H

InChI key

PIWKPBJCKXDKJR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoflurane EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Isoflurane is a tandem pore potassium channel activator.
Isoflurane is a tandem pore potassium channel activator. It is also a very widely used anesthetic for animal research and for in vitro studies on anesthesia mechanisms.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanlin Jia et al.
Nature communications, 5, 4387-4387 (2014-07-19)
Injury to lung epithelial cells has a role in multiple lung diseases. We previously identified mitsugumin 53 (MG53) as a component of the cell membrane repair machinery in striated muscle cells. Here we show that MG53 also has a physiological
Oliver Grottke et al.
Critical care (London, England), 18(1), R27-R27 (2014-02-07)
New oral anticoagulants are effective alternatives to warfarin. However, no specific reversal agents are available for life-threatening bleeding or emergency surgery. Using a porcine model of trauma, this study assessed the ability of prothrombin complex concentrate (PCC), activated PCC (aPCC)
David A X Nayagam et al.
PloS one, 9(5), e97182-e97182 (2014-05-24)
To assess the safety and efficacy of chronic electrical stimulation of the retina with a suprachoroidal visual prosthesis. Seven normally-sighted feline subjects were implanted for 96-143 days with a suprachoroidal electrode array and six were chronically stimulated for 70-105 days
Qiaoya Lin et al.
Small (Weinheim an der Bergstrasse, Germany), 10(15), 3072-3082 (2014-04-08)
The abilities to deliver siRNA to its intended action site and assess the delivery efficiency are challenges for current RNAi therapy, where effective siRNA delivery will join force with patient genetic profiling to achieve optimal treatment outcome. Imaging could become
Zareen Amtul et al.
Neurobiology of aging, 35(7), 1605-1614 (2014-02-05)
Numerous clinical and epidemiological reports indicate that patients with history of vascular illness such as stroke are more likely to develop dementia as the clinical manifestation of Alzheimer's disease. However, there are little data regarding the pathologic mechanisms that link

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service