Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000762

Fluorouracil impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Barbituric acid, 2,4,6-Trihydroxypyrimidine, Malonylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O3
CAS Number:
Molecular Weight:
128.09
Beilstein:
120502
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluorouracil

manufacturer/tradename

EDQM

mp

248-252 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1CC(=O)NC(=O)N1

InChI

1S/C4H4N2O3/c7-2-1-3(8)6-4(9)5-2/h1H2,(H2,5,6,7,8,9)

InChI key

HNYOPLTXPVRDBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluorouracil impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

302.0 °F - closed cup

Flash Point(C)

150.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayman K Hamouda et al.
Molecular pharmacology, 85(5), 735-746 (2014-02-25)
At concentrations that produce anesthesia, many barbituric acid derivatives act as positive allosteric modulators of inhibitory GABAA receptors (GABAARs) and inhibitors of excitatory nicotinic acetylcholine receptors (nAChRs). Recent research on [(3)H]R-mTFD-MPAB ([(3)H]R-5-allyl-1-methyl-5-(m-trifluoromethyldiazirinylphenyl)barbituric acid), a photoreactive barbiturate that is a potent
Roosmarijn E Vandenbroucke et al.
Nature reviews. Drug discovery, 13(12), 904-927 (2014-11-08)
Matrix metalloproteinases (MMPs) are zinc-dependent endopeptidases that form a family of 24 members in mammals. Evidence of the pathological roles of MMPs in various diseases, combined with their druggability, has made them attractive therapeutic targets. Initial drug discovery efforts focused
Rémi Legay et al.
Journal of pharmaceutical and biomedical analysis, 98, 446-462 (2014-07-20)
The purpose of the study was to investigate the degradation pathway of 5-fluorouracil (FU) in the situation of commercial formulations for clinical use, namely FU dissolved in sodium hydroxide (NaOH) solutions or Tris buffer at pH 8.5-9. Combination of data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service