Skip to Content
MilliporeSigma
All Photos(1)

Documents

Y0000683

Phenylephrine hydrochloride for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(R)-(−)-Phenylephrine hydrochloride, (R)-(−)-1-(3-Hydroxyphenyl)-2-methylaminoethanol hydrochloride, (R)-(−)-3-Hydroxy-α-(methylaminomethyl)benzyl alcohol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH(CH2NHCH3)OH·HCl
CAS Number:
Molecular Weight:
203.67
Beilstein:
4158948
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenylephrine

manufacturer/tradename

EDQM

mp

143-145 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC1=CC([C@H](CNC)O)=CC=C1.Cl

InChI

1S/C9H13NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,9-12H,6H2,1H3;1H/t9-;/m0./s1

InChI key

OCYSGIYOVXAGKQ-FVGYRXGTSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenylephrine hydrochloride for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dale A Currigan et al.
Anesthesiology, 121(5), 930-936 (2014-09-10)
Vasopressor drugs, commonly used to treat systemic hypotension and maintain organ perfusion, may also induce regional vasoconstriction in specialized vascular beds such as the lung. An increase in pulmonary vascular tone may adversely affect patients with pulmonary hypertension or right
C A Boly et al.
British journal of anaesthesia, 111(5), 750-758 (2013-07-11)
Left-ventricular end-systolic elastance (Ees) is an index of cardiac contractility, but the invasive nature of its assessment has limited perioperative application. We explored the feasibility of a minimally invasive method of Ees estimation for perioperative assessment of cardiac function and
Stephanie W Watts et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(6), 1320-1328 (2013-04-06)
Obesity and hypertension are comorbid in epidemic proportion, yet their biological connection is largely a mystery. The peptide chemerin is a candidate for connecting fat deposits around the blood vessel (perivascular adipose tissue) to arterial contraction. We presently tested the
Heather Mak et al.
Ophthalmology, 120(12), 2517-2524 (2013-07-16)
To measure iris volume and anterior segment parameters using a swept-source anterior segment optical coherence tomography (OCT) and investigate factors associated with iris volume and iris volume change after pupil dilation in eyes with open angles and angle closure. Cross-sectional
Jacky W Y Lee et al.
Medicine, 94(8), e567-e567 (2015-02-26)
To investigate the association between macular thickness and peripapillary retinal nerve fiber layer (RNFL) thickness in Chinese children. This cross-sectional study recruited consecutive cases of healthy pediatric subjects aged 4 to 18 from Caritas Medical Centre in Hong Kong Special

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service