Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000434

Boldine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,9-Dihydroxy-1,10-dimethoxyaporphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
Beilstein:
94036
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

boldine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1cc-2c(CC3N(C)CCc4cc(O)c(OC)c-2c34)cc1O

InChI

1S/C19H21NO4/c1-20-5-4-10-7-15(22)19(24-3)18-12-9-16(23-2)14(21)8-11(12)6-13(20)17(10)18/h7-9,13,21-22H,4-6H2,1-3H3/t13-/m0/s1

InChI key

LZJRNLRASBVRRX-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Boldine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Darina Muthna et al.
Natural product communications, 8(12), 1797-1800 (2014-02-22)
Plant and folk medicine represent nowadays a source of either new therapeutic substances or substrates for drug synthesis. One such promising group for possible further exploitation is the family of aporphine alkaloids containing boldine and related compounds. In this mini-review
Peter O'Brien et al.
Chemico-biological interactions, 159(1), 1-17 (2005-10-14)
The increasing recognition of the participation of free radical-mediated oxidative events in the initiation and/or progression of cardiovascular, tumoural, inflammatory and neurodegenerative disorders, has given rise to the search for new antioxidant molecules. An important source of such molecules has
H Speisky et al.
Pharmacological research, 29(1), 1-12 (1994-01-01)
Boldo (Peumus boldus Mol.), a Chilean tree traditionally employed in folk medicine and recognized as a herbal remedy in a number of pharmacopoeias, mainly for the treatment of liver ailments, has recently been the subject of increasing attention. Boldine, in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service