Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000101

Orphenadrine citrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Orphenadrine citrate salt, β-Dimethylaminoethyl 2-methylbenzhydryl ether citrate salt, o-Methyldiphenhydramine citrate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23NO · C6H8O7
CAS Number:
Molecular Weight:
461.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

orphenadrine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CC(CC(O)=O)C(O)=O.CN(C)CCOC(c1ccccc1)c2ccccc2C

InChI

1S/C18H23NO.C6H8O6/c1-15-9-7-8-12-17(15)18(20-14-13-19(2)3)16-10-5-4-6-11-16;7-4(8)1-3(6(11)12)2-5(9)10/h4-12,18H,13-14H2,1-3H3;3H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

XLSFUJUURNFSCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Orphenadrine citrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Muscarinic receptor antagonist; H1 histamine receptor antagonist; muscle relaxant. Orphenadrine has also been reported to inhibit the noradrenergic transporter and to block the NMDA receptor ion channel.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohamed I Walash et al.
Chemistry Central journal, 5, 60-60 (2011-10-11)
A simple, sensitive and rapid spectrophotometric method was developed and validated for the determination of two skeletal muscle relaxants namely, tizanidine hydrochloride (I) and orphenadrine citrate (II) in pharmaceutical formulations. The proposed method is based on the formation of a
Letizzia Dall'Agnol et al.
The journal of pain, 15(8), 845-855 (2014-05-29)
Chronic myofascial pain syndrome has been related to defective descending inhibitory systems. Twenty-four females aged 19 to 65 years with chronic myofascial pain syndrome were randomized to receive 10 sessions of repetitive transcranial magnetic stimulation (rTMS) (n = 12) at
Jean-François Desaphy et al.
Experimental neurology, 255, 96-102 (2014-03-13)
Although the sodium channel blocker mexiletine is considered the first-line drug in myotonia, some patients experiment adverse effects, while others do not gain any benefit. Other antimyotonic drugs are thus needed to offer mexiletine alternatives. In the present study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service