Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Y0000047

Levamisole hydrochloride for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Tetramisole hydrochloride, (S)-(−)-6-Phenyl-2,3,5,6-tetrahydroimidazo[2,1-b]thiazole hydrochloride, L(−)-2,3,5,6-Tetrahydro-6-phenylimidazo[2,1-b]thiazole hydrochloride, Levamisole hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2S · HCl
CAS Number:
Molecular Weight:
240.75
Beilstein:
4358988
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levamisole

manufacturer/tradename

EDQM

mp

228-230 °C

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.C1CN2C[C@@H](N=C2S1)c3ccccc3

InChI

1S/C11H12N2S.ClH/c1-2-4-9(5-3-1)10-8-13-6-7-14-11(13)12-10;/h1-5,10H,6-8H2;1H/t10-;/m1./s1

InChI key

LAZPBGZRMVRFKY-HNCPQSOCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levamisole hydrochloride for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Shows both immunostimulant and immunosuppressant effects, depending on several controllable factors. Very effective in treatment of ascariasis (hookworm infestation). Useful in chemotherapy of colorectal cancers, possibly due to its stimulation of IL-1 production and direct activation of macrophages.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cara Hennings et al.
Journal of the American Academy of Dermatology, 69(1), 135-142 (2013-02-13)
We review the most common systemic and cutaneous signs of heroin, cocaine, methamphetamine, Ecstasy, and marijuana use. We also provide an overview of the skin and soft-tissue infections frequently found in intravenous drug users and the effects of the adulterants
Barbara Deracinois et al.
PloS one, 7(10), e48428-e48428 (2012-11-03)
Although the physiological properties of the blood-brain barrier (BBB) are relatively well known, the phenotype of the component brain capillary endothelial cells (BCECs) has yet to be described in detail. Likewise, the molecular mechanisms that govern the establishment and maintenance
Anders N Hansen et al.
PloS one, 7(9), e45405-e45405 (2012-10-02)
Inhibition of angiogenesis is a promising addition to current cancer treatment strategies. Neutralization of vascular endothelial growth factor by monoclonal antibodies is clinically effective but may cause side effects due to thrombosis. Low molecular weight angiogenesis inhibitors are currently less
Bashir A Lone et al.
Tropical animal health and production, 45(3), 743-749 (2012-10-16)
The aim of this study was to evaluate the anthelmintic and antimicrobial efficacy of Euphorbia helioscopia crude extracts. A worm motility inhibition assay and egg hatch assay were used for in vitro study, and a faecal egg count reduction assay
Lauren Strazzula et al.
Journal of the American Academy of Dermatology, 69(6), 954-959 (2013-10-01)
Levamisole is present as a contaminant or additive in most cocaine sold in the United States. Cases of agranulocytosis attributed to levamisole-tainted cocaine have been widely described. A vasculopathic reaction to levamisole has also been reported; however, diagnostic features such as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service