Skip to Content
MilliporeSigma
All Photos(1)

Documents

W421350

Sigma-Aldrich

Farnesyl acetate

≥95%, stabilized, FG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28O2
CAS Number:
Molecular Weight:
264.40
FEMA Number:
4213
Beilstein:
1784823
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.818
NACRES:
NA.21

grade

FG
Halal
Kosher

Quality Level

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

contains

synthetic α-tocopherol as stabilizer

density

0.91 g/mL at 20 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

green; floral; rose

SMILES string

CC(=O)OC\C=C(/C)CC\C=C(/C)CC\C=C(\C)C

InChI

1S/C17H28O2/c1-14(2)8-6-9-15(3)10-7-11-16(4)12-13-19-17(5)18/h8,10,12H,6-7,9,11,13H2,1-5H3/b15-10+,16-12+

InChI key

ZGIGZINMAOQWLX-NCZFFCEISA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service