Skip to Content
MilliporeSigma
All Photos(2)

Documents

W417170

Sigma-Aldrich

Methyl N-formylanthranilate

≥98%, FG

Synonym(s):

Methyl 2-formamidobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO3
CAS Number:
Molecular Weight:
179.17
FEMA Number:
4171
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.650
NACRES:
NA.21

grade

FG
Fragrance grade

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥98%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

grape; fruity

InChI

1S/C9H9NO3/c1-13-9(12)7-4-2-3-5-8(7)10-6-11/h2-6H,1H3,(H,10,11)

InChI key

HRNPZFOYXWWMFL-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service