Skip to Content

STS0004

TERGITOL TMN-100X solution

Synonym(s):

Branched secondary alcohol ethoxylate 10.1 EO, Polyethylene glycol trimethylnonyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

laboratory grade

Quality Segment

description

non-ionic

form

liquid

mol wt

570 g/mol by calculation

technique(s)

HPLC: suitable, LC/MS: suitable

CMC

830 ppm (25°C)

viscosity

88 cP(25 °C)

transition temp

pour point -6 °C, cloud point 65 °C (1 wt% actives aq solution)

solubility

water: freely soluble at 20 °C (visual)

density

1.025 g/mL at 20 °C

HLB

14.1

SMILES string

O(C(CC(CC(C)C)C)CC(C)C)CCO

InChI

1S/C14H30O2/c1-11(2)8-13(5)10-14(9-12(3)4)16-7-6-15/h11-15H,6-10H2,1-5H3

InChI key

VKKFWRYCMZBXIG-UHFFFAOYSA-N

General description

This product is a 90% aqueous solution of a branched secondary alcohol ethoxylate with 10.1 ethylene oxide (EO) units.

Application

TERGITOL TMN-100X can be used as:
  • A foaming agent in the study of surfactant effect on glass-ceramic biomaterials (bioactive glasses).
  • A surfactant in the preparation of nickel-graphene oxide composite coatings.

Features and Benefits

  • Superior wetting
  • Penetrant & dispersant
  • Narrow gel range
  • Effective emulsifier

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow


Pictograms

Corrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class

10 - Combustible liquids

WGK

WGK 2

Flash Point (°F)

>230.0 °F

Flash Point (°C)

> 110 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library