Skip to Content
MilliporeSigma
All Photos(1)

Documents

SMB01381

Sigma-Aldrich

SAICAriboside

≥95% (HPLC), from synthetic, solid

Synonym(s):

N-Succinyl-5-aminoimidazole-4-carboxamide Ribose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18N4O9
CAS Number:
Molecular Weight:
374.30
MDL number:
UNSPSC Code:
12352211
UNSPSC Code:
12352200
NACRES:
NA.26

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

solid

color

white to beige

mp

130-135 °C

storage temp.

2-8°C

SMILES string

NC1=C(C(N[C@H](C(O)=O)CC(O)=O)=O)N=CN1[C@H]2[C@H](O)[C@H](O)[C@@H](CO)O2

General description

SAICAR (a ribotide) can lose its phosphate group leading to the appearance of a riboside known as succinylaminoimidazolecarboxamide riboside (SAICAriboside or SAICAr) in cerebrospinal fluid, in urine, and, to a lesser extent, in plasma. SAICAriboside is characteristic of ADSL, a heritable deficiency of the enzyme adenylosuccinate lyase (ASL or adenylosuccinase) responsible for metabolizing SAICAR (SZMP) to AICAR (ZMP) and adenylosuccinate (SAMP) to AMP. ASL deficiency causes increased SAICAR & SAMP, and their corresponding rephosphorylated products SAICAr & succinyladenosine (S-Ado).

Storage and Stability

Heat sensitive

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of purinosome assembly in cell-based model systems with de novo purine synthesis and salvage pathway deficiencies
Baresova, et al.
PLoS ONE, 13, e0201432-e0201432 (2018)
Roxane Marsac et al.
Genetics, 211(4), 1297-1313 (2019-02-01)
Purine homeostasis is ensured through a metabolic network widely conserved from prokaryotes to humans. Purines can either be synthesized de novo, reused, or produced by interconversion of extant metabolites using the so-called recycling pathway. Although thoroughly characterized in microorganisms, such
Delphine C Douillet et al.
The Journal of biological chemistry, 294(3), 805-815 (2018-11-28)
5-Aminoimidazole-4-carboxamide 1-β-d-ribofuranoside (AICAR, or acadesine) is a precursor of the monophosphate derivative 5-amino-4-imidazole carboxamide ribonucleoside 5'-phosphate (ZMP), an intermediate in de novo purine biosynthesis. AICAR proved to have promising anti-proliferative properties, although the molecular basis of its toxicity is poorly
Mass spectrometric analysis of purine de novo biosynthesis intermediates
Madrova, et al.
PLoS ONE, 13, e0208947-e0208947 (2018)
A mild form of adenylosuccinate lyase deficiency in absence of typical brain MRI features diagnosed by whole exome sequencing
Macchiaiolo, et al.
Italian Journal of Pediatrics, 43, 65-65 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service