Skip to Content
MilliporeSigma
All Photos(1)

Documents

S2150000

Sulfamethoxypyridazine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N1-(6-Methoxypyridazin-3-yl)sulfanilamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N4O3S
CAS Number:
Molecular Weight:
280.30
Beilstein:
277076
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sulfamethoxypyridazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(NS(=O)(=O)c2ccc(N)cc2)nn1

InChI

1S/C11H12N4O3S/c1-18-11-7-6-10(13-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,15)

InChI key

VLYWMPOKSSWJAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sulfamethoxypyridazine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[SIDE-EFFECTS OF SULFAMETHOXYPYRIDAZINE].
K DVORAK
Lekarska veda v zahranici, 41, 135-142 (1963-07-08)
Joseph M Storey et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 972, 38-47 (2014-10-14)
A liquid chromatography-tandem mass spectrometry (LC-MS/MS) screening method is described for the detection and identification of 26 veterinary drugs in fish and other aquaculture products. The analytes include: 13 sulfonamides, trimethoprim, 3 fluoroquinolones, 3 quinolones, 3 triphenylmethane dyes, 2 leuco
Hui-zhen Wu et al.
Analytical and bioanalytical chemistry, 407(13), 3545-3554 (2015-03-04)
An in-line matrix cleanup method was used for the simultaneous extraction of 15 sulfonamides and two metabolites from manure samples. The ultrasound/microwave-assisted extraction (UMAE) combined with solid-liquid-solid dispersive extraction (SLSDE) procedure provides a simple sample preparation approach for the processing
Yong-Gang Zhao et al.
Journal of chromatography. A, 1345, 17-28 (2014-05-02)
A novel, simple and sensitive method was developed for the simultaneous determination of 22 sulfonamides (SAs) in chicken breast muscle by using the dispersive micro-solid-phase extraction (d-μ-SPE) procedure combined with ultra-fast liquid chromatography-tandem quadrupole mass spectrometry (UFLC-MS/MS). The excellent core-shell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service