Skip to Content
MilliporeSigma

RTC000095

Methyl Isobutyl Ketone

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
605399
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

grade

certified reference material, pharmaceutical secondary standard

agency

traceable to USP 1430203

vapor density

3.5 (vs air)

CofA

current certificate can be downloaded

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

packaging

pkg of 20 mL

refractive index

n20/D 1.395 (lit.)

bp

117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

application(s)

pharmaceutical small molecule

storage temp.

2-30°C

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Analysis Note

These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB9803 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.


Still not finding the right product?

Explore all of our products under Methyl Isobutyl Ketone


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

57.2 °F

flash_point_c

14 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library