Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

RTC000068

Supelco

Sodium azide solution

0.05% solution

Synonym(s):

NaN3 0.05% Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352125
NACRES:
NA.24

Product Name

Sodium Azide 0.05% Solution,

CofA

current certificate can be downloaded

Quality Level

packaging

bottle of 1 L

concentration

in H2O

format

single component solution

storage temp.

2-8°C

General description

An analytical standard with an assigned value of 0.05%, measured based on purity determination by mass balance and gravimetry.
The certified reference material (CRM) standard is designed, manufactured and tested according to ISO/IEC 17025:2017 and ISO 17034:2016.

Application

Intended for R&D and analytical use only.

Features and Benefits

  • certified purity value of 0.0500 ± 0.0020 is metrologically traceable to National Institute Standards and Technology Standard Reference Materials (NIST SRMs)
  • Details of homogeneity studies and product expiration are provided in the certificate of analysis (CoA)
  • Solvent/Matrix: Type 1 water
  • Packaging: high-density poly ethylene (HDPE) bottle

Analysis Note

This is a solution in Water

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service