Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR3609

Supelco

N-Nitrosoethylisopropylamine (NEIPA / NIPEA)

certified reference material, pharmaceutical secondary standard

Synonym(s):

N-Ethyl-N-nitroso-2-propanamine, EIPNA, NEIPA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O
CAS Number:
Molecular Weight:
116.16
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1466685

form

methanol solution

CofA

current certificate can be downloaded

packaging

pkg of 1 mL

application(s)

pharmaceutical

storage temp.

-10 to -25°C

SMILES string

N(N=O)(C(C)C)CC

InChI

1S/C5H12N2O/c1-4-7(6-8)5(2)3/h5H,4H2,1-3H3

InChI key

VGGZTNNNXAUZLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

N-Nitrosoethylisopropylamine pharmaceutical secondary reference standard can be used for the determination of NEIPA impurity in valsartan products, via gas chromatography-tandem mass spectrometry.

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Solvent

Description

    1000 ug/mL solution in methanol

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

150.6 °F - (calculated)

Flash Point(C)

65.9 °C - (calculated)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development of GC-MS/MS Method for Simultaneous Estimation of Four Nitrosoamine Genotoxic Impurities in Valsartan
Rao T, et al.
Turkish Journal of Pharmaceutical Sciences, 19, 455-455 (2022)

Articles

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Discover the efficient separation six nitrosamine impurities with high reproducibility and precise detection to ensure accurate analysis for regulatory compliance.

Related Content

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Accurately detect and quantify trace nitrosamines (NDMA, NDEA, NEIPA, NDIPA, NDBA, etc.) in pharmaceutical drugs using our complete product portfolio and application guides for LC-MS and GC-MS. Order high-quality reference standards, columns, filters & more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service