Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1903

Supelco

Citalopram Related Compound D

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Citalopram Related Compound D, 1-(4-Fluorophenyl)-1-(3-methylaminopropyl)-1,3-dihydroisobenzofuran-5-carbonitrile hydrochloride, 1-(4-Fluorophenyl)-1-(3-methylamino)propyl)-1,3-dihydroisobenzofuran-5-carbonitrile hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H19FN2O · HCl
CAS Number:
Molecular Weight:
346.83
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1134277

API family

citalopram

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Fc1ccc(cc1)C2(OCc3c2ccc(c3)C#N)CCCNC.Br

InChI

1S/C19H19FN2O.BrH/c1-22-10-2-9-19(16-4-6-17(20)7-5-16)18-8-3-14(12-21)11-15(18)13-23-19;/h3-8,11,22H,2,9-10,13H2,1H3;1H

InChI key

DJCAYVHVPUEMQJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Citalopram Related Compound D is a related impurity found in the drug citalopram. Citalopram is a racemic bicyclic phthalane derivative and is a potent anti-depressant drug, that acts by inhibition of serotonin uptake.

Application

Citalopram Related Compound D may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB0345 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development and optimization of an HPLC analysis of citalopram and its four nonchiral impurities using experimental design methodology
Tadi T, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 95(3), 733-743 (2012)
Semi-preparative isolation and structural elucidation of an impurity in citalopram by LC/MS/MS
Raman B, et al.
Journal of Pharmaceutical and Biomedical Analysis, 50(3), 377-383 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service