Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1873

Supelco

Atorvastatin Related Compound H

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

ATORVASTATIN RELATED COMPOUND H, Lactone Impurity, 5-(4-Fluorophenyl)-1-{2-[(2R, 4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethyl}-2-isopropyl-N,4-diphenyl-1H-pyrrole-3-carboxamide, (Lactone Impurity, 5-(4-Fluorophenyl)-1-{2-[(2R,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-
yl]ethyl}-2-isopropyl-N,4-diphenyl-1H-pyrrole-3-carboxamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H33FN2O4
CAS Number:
Molecular Weight:
540.62
UNSPSC Code:
12161503
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1044582

API family

atorvastatin

CofA

current certificate can be downloaded

packaging

pkg of 30 mg

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

InChI

1S/C33H33FN2O4/c1-21(2)31-30(33(39)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-27-19-26(37)20-28(38)40-27/h3-16,21,26-27,37H,17-20H2,1-2H3,(H,35,39)/t26-,27-/m1/s1

InChI key

OUCSEDFVYPBLLF-KAYWLYCHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB1039 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A validated capillary electrophoresis method for simultaneous determination of ezetimibe and atorvastatin in pharmaceutical formulations
Alshehri MM
Saudi Pharmaceutical Journal : SPJ : The Official Publication of the Saudi Pharmaceutical Society, 20(2), 143-148 (2012)
Stability indicating method development for simultaneous estimation of ezetimibe and atorvastatin in pharmaceutical formulations by RP-HPLC
PK, et al.
Pharmaceutica Analytica Acta, 3(6), 1-6 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service