Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1817

Supelco

Furosemide Related Compound A

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-Chloro-4-furfurylamino-5-sulfamoylbenzoic acid, 2-CHLORO-4-N-FURFURYLAMINO -5-SULFAMOYLBENZOIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11ClN2O5S
CAS Number:
Molecular Weight:
330.74
Beilstein:
1297887
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. F0702000
traceable to USP 1287020

CofA

current certificate can be downloaded

packaging

pkg of 100 mg

mp

197 °C (dec.) (lit.)

application(s)

pharmaceutical

format

neat

storage temp.

2-30°C

SMILES string

OC(C1=C(Cl)C=C(NCC2=CC=CO2)C(S(N)(=O)=O)=C1)=O

InChI

1S/C12H11ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)11(21(14,18)19)4-8(9)12(16)17/h1-5,15H,6H2,(H,16,17)(H2,14,18,19)

InChI key

UXOOVYKVEXGCSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application


  • Furosemide Impurity A for Quality Control: Furosemide Related Compound A serves as a crucial standard in pharmaceutical quality control, ensuring the purity and safety of furosemide as an active pharmaceutical ingredient. This impurity standard is essential for regulatory compliance and batch-to-batch consistency in drug manufacturing (Abasilim et al., 2023).

  • Research on Drug Resistance: Furosemide Related Compound A is utilized in genomic research to identify potential drug resistance mechanisms. This research aids in understanding how bacterial pathogens develop resistance to drugs like clarithromycin and rifampicin, which is crucial for developing new therapeutic strategies (Mofidifar et al., 2024).

  • Nanotechnology in Drug Delivery: High-purity Furosemide Related Compound A is used in the preparation and characterization of novel drug delivery systems, such as nanoparticles designed to enhance the bioavailability and efficacy of pharmaceutical compounds. This research highlights its application in improving drug formulations and targeting specific disease sites (Zeng et al., 2024).

  • Impact on Bone Health: Research involving Furosemide Related Compound A explores its effects on bone health, particularly in how it influences vitamin D metabolism and bone density. This is vital for developing treatments that mitigate bone density loss associated with long-term use of diuretics like furosemide (Melanie KF et al., 2024).

  • Nutritional Studies in Cancer Therapy: Furosemide Related Compound A is also examined for its role in nutritional interventions in cancer therapy, particularly its impact on the nutritional status of patients undergoing surgery for colorectal cancer. These studies are crucial for understanding how nutritional support can enhance recovery and treatment outcomes in cancer patients (Ural et al., 2024).

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC2891 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service