Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1725

Supelco

Glimepiride cis-Isomer (Glimepiride Related Compound A)

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

3-Ethyl-2,5-dihydro-4-methyl-N-[2-[4-[[[[(cis-4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-oxo-1H-pyrrole-1-carboxamide, 1-[[p-[2-(3-Ethyl-4-methyl-2-oxo-3-pyrroline-1-carboxamido)ethyl]phenyl]sulfonyl]-3-(cis-4-methylcyclohexyl)urea, Glimepiride cis-isomer

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34N4O5S
CAS Number:
Molecular Weight:
490.62
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1292314

API family

glimepiride

CofA

current certificate can be downloaded

packaging

pkg of 40 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(NC(=O)N[C@@H]3CC[C@@H](CC3)C)c1ccc(cc1)CCNC(=O)N2CC(=C(C2=O)CC)C

InChI

1S/C24H34N4O5S/c1-4-21-17(3)15-28(22(21)29)24(31)25-14-13-18-7-11-20(12-8-18)34(32,33)27-23(30)26-19-9-5-16(2)6-10-19/h7-8,11-12,16,19H,4-6,9-10,13-15H2,1-3H3,(H,25,31)(H2,26,27,30)/t16-,19+

InChI key

WIGIZIANZCJQQY-UWUNEBHHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glimepiride cis-Isomer (Glimepiride Related Compound A) is an impurity of glimepiride. Glimepiride is a third generation, medium-to-long acting sulphonylurea compound used as an oral antidiabetic drug. Its mode of action involves the lowering of blood glucose by stimulating the release of insulin from functioning pancreatic beta cells.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Glimepiride cis-Isomer (Glimepiride Related Compound A) may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7400 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glimepiride
USP42-NF37: United States Pharmacopeia and National Formulary
United States Pharmacopeia/National Formulary, 42(6), 2062-2062 (2018)
LC determination of glimepiride and its related impurities
Khan MA, et al.
Journal of Pharmaceutical and Biomedical Analysis, 39(5), 928-943 (2005)
RP-HPLC analytical method development and validation for simultaneous estimation of three drugs: Glimepiride, pioglitazone, and metformin and its pharmaceutical dosage forms
Nirupa G and Tripathi UM
Journal of Chemistry, 2013(5), 550-559 (2012)
Rudy Bonfilio et al.
Therapeutic drug monitoring, 32(5), 550-559 (2010-08-26)
Glimepiride is an oral antidiabetic drug in the sulfonylurea class, which is widely used in treatment of Type 2 diabetes and it is currently available in more than 60 countries worldwide. As a result of the importance of this oral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service