Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1603

Supelco

Montelukast Sodium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Montelukast sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H35ClNO3S·Na
CAS Number:
Molecular Weight:
608.17
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001434
traceable to USP 1446859

API family

montelukast

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].S([C@H](CCc5c(cccc5)C(O)(C)C)c2cc(ccc2)\C=C\c3nc4c(cc3)ccc(c4)Cl)CC1(CC1)CC(=O)[O-]

InChI

1S/C35H36ClNO3S.Na/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(41-23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29;/h3-12,14-16,20-21,32,40H,13,17-19,22-23H2,1-2H3,(H,38,39);/q;+1/p-1/b15-10+;/t32-;/m1./s1

InChI key

LBFBRXGCXUHRJY-HKHDRNBDSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Montelukast Sodium is an orally administered leukotriene receptor antagonist drug that acts by inhibiting the cysteinyl leukotriene (CysLT1) receptor. It is effective against leukotrienes responsible for bronchoconstriction and inflammation of the airways leading to asthma problems.

Application

Montelukast Sodium may be used as a pharmaceutical reference standard for the determination of the analyte in bulk drug and pharmaceutical formulations by chromatography and spectrophotometric techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3617 in the slot below. This is an example certificate only and may not be the lot that you receive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric method for quantitative determination of montelukast in bulk, pharmaceutical formulations and human serum
Arayne MS, et al.
Journal of Analytical Chemistry, 64(7), 690-695 (2009)
Montelukast Sodium
USP42-NF37
United States Pharmacopeia/National Formulary, 37(2), 2974-2974 (2018)
HPTLC determination of montelukast sodium in bulk drug and in pharmaceutical preparations
Sane R, et al.
J. Planar Chromatogr., 17(1), 75-78 (2004)
Santa Mandal et al.
Inhalation toxicology, 32(7), 299-310 (2020-07-01)
To explore the therapeutic role of a single dose combination of montelukast (MON) and dexamethasone (DXM) through intra-peritoneal route against paraquat (PQ)-intoxicated experimental Wistar rats. In vivo the survival rate was investigated following the administration of both MON and DXM
Sheng-Hua Wu et al.
Pediatrics international : official journal of the Japan Pediatric Society, 56(3), 315-322 (2013-12-05)
Previous studies suggested that leukotrienes (LT) were involved in the pathogenesis of Henoch-Schönlein purpura (HSP). This study investigated the efficacy of an add-on therapy with montelukast in the treatment of HSP. In this four-center, double-blind, placebo-controlled, parallel paired comparative study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service