Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHR1585

Supelco

Esomeprazole Magnesium

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-Omeprazole magnesium trihydrate, 5-Methoxy-2-{(S)-[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl] sulfinyl} benzimidazole magnesium trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H36MgN6O6S2 · 3H2O
CAS Number:
Molecular Weight:
767.17
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001028
traceable to USP 1249789

API family

omeprazole

CofA

current certificate can be downloaded

packaging

ampule of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

[Mg+2].[S](=O)(Cc6ncc(c(c6C)OC)C)c4[n-]c5c(n4)cc(cc5)OC.[S](=O)(Cc3ncc(c(c3C)OC)C)c1[n-]c2c(n1)cc(cc2)OC.O.O.O

InChI

1S/2C17H18N3O3S.Mg.3H2O/c2*1-10-8-18-15(11(2)16(10)23-4)9-24(21)17-19-13-6-5-12(22-3)7-14(13)20-17;;;;/h2*5-8H,9H2,1-4H3;;3*1H2/q2*-1;+2;;;

InChI key

VEVZQDGATGBLIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Esomeprazole Magnesium is theS-isomer of omeprazole and is widely used for the treatment of acid related diseases. It is the first single optical isomer proton pump inhibitor and is found to be better over omeprazole in terms of pharmacokinetic characteristics and acid suppression.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Esomeprazole Magnesium may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using UV-Vis spectroscopy technique and spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1677 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric Determination of Esomeprazole Magnesium in Commercial Tablets Using 5?Sulfosalicylic Acid and N?Bromosuccinimide
Rahman N, et al.
J. Chin. Chem. Soc., 55(3), 557-566 (2008)
Development and validation of analytical methods for Simultaneous estimation of domperidone and esomeprazole Magnesium in bulk and in pharmaceutical formulations Using UV-Visible spectroscopy
BPD, et al.
Development, 6(5), 1501-1508 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service