Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1015

Supelco

Diphenhydramine hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

DP, DPH, 2-Diphenylmethoxy-N,N-dimethylethylamine hydrochloride, N-(2-Diphenylmethoxyethyl)-N,N-dimethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO · HCl
CAS Number:
Molecular Weight:
291.82
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 896
traceable to Ph. Eur. D2000000
traceable to USP 1218005

API family

diphenhydramine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl[H].CN(C)CCOC(c1ccccc1)c2ccccc2

InChI

1S/C17H21NO.ClH/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;/h3-12,17H,13-14H2,1-2H3;1H

InChI key

PCHPORCSPXIHLZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Diphenhydramine hydrochloride may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7790 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimization and validation of a method for the determination of caffeine, 8-chlorotheophylline and diphenhydramine by isocratic high-performance liquid chromatography: Stress test for stability evaluation
Barbas C, et al.
Journal of Chromatography A, 870(1), 97-103 (2000)
Quantitative analysis of liquid formulations using FT-Raman spectroscopy and HPLC: The case of diphenhydramine hydrochloride in [TM="Benadryl"]
Orkoula MG, et al.
Journal of Pharmaceutical and Biomedical Analysis, 41(4), 1406-1411 (2006)
Development and validation of a capillary electrophoresis method for the determination of codeine, diphenhydramine, ephedrine and noscapine in pharmaceuticals
Gomez MR, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 60(1), 85-90 (2005)
Analysis of pharmaceutical preparations containing antihistamine drugs by micellar liquid chromatography
Martinez-Algaba C, et al.
Journal of Pharmaceutical and Biomedical Analysis, 40(2), 312-321 (2006)
Eugene W Schwilke et al.
Journal of forensic sciences, 51(5), 1191-1198 (2006-10-05)
We have previously reported on patterns of drug and alcohol use in fatally injured drivers in Washington State. Here we revisit that population to examine how drug use patterns have changed in the intervening 9 years. Blood and serum specimens

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service