Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHL89675

Geniposide

phyproof® Reference Substance

Synonym(s):

(1S,4αS,7αS)- 1-(β-D-Glucopyranosyloxy)-1,4a,5,7a-tetrahydro-7-(hydroxymethyl)- cyclopenta[c]pyran-4-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24O10
CAS Number:
Molecular Weight:
388.37
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H]3[C@@H]1CC=C3CO

InChI

1S/C17H24O10/c1-24-15(23)9-6-25-16(11-7(4-18)2-3-8(9)11)27-17-14(22)13(21)12(20)10(5-19)26-17/h2,6,8,10-14,16-22H,3-5H2,1H3/t8-,10-,11-,12-,13+,14-,16+,17+/m1/s1

InChI key

IBFYXTRXDNAPMM-BVTMAQQCSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Geniposide is an iridoid glycoside with a variety of biological activities including neuroprotective, anti-diabetic, antiproliferative, and antioxidative activity. Geniposide has been shown to regulate Nrf2 translocation and upregulate downstream antioxidant protein HO-1 expression through the PI3K/Akt signaling pathway, which may account for its neuroprotective and antioxidative activity. Glucagon-like peptide 1 (GLP-1) receptor may also play a critical role in geniposide induced HO-1 expression.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Analyze Xiaoer Chiqiao Qingre granules for geniposide and forsythin using Discovery® HS C18 column and Supelclean™ LC-Alumina-N SPE.

Analyze Xiaoer Chiqiao Qingre granules for geniposide and forsythin using Discovery® HS C18 column and Supelclean™ LC-Alumina-N SPE.

Analyze Xiaoer Chiqiao Qingre granules for geniposide and forsythin using Discovery® HS C18 column and Supelclean™ LC-Alumina-N SPE.

Analyze Xiaoer Chiqiao Qingre granules for geniposide and forsythin using Discovery® HS C18 column and Supelclean™ LC-Alumina-N SPE.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service