Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL82614

Ophiopogonin D

phyproof® Reference Substance

Synonym(s):

(1β,3β,25R)-3-Hydroxyspirost-5-en-1-yl O-6-deoxy-α-L-mannopyranosyl-(1→2)-O-[β-D-xylopyranosyl-(1→3)]-6-deoxy-β-D-galactopyranoside, 25(R)-Ruscogenin 1-O-[α-L-rhamnopyranosyl-(1→2)][β-D-xylopyranosyl-(1→3)]-β-D-fucopyranoside, Deacetylophiopogonin C, OJV-V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H70O16
CAS Number:
Molecular Weight:
855.02
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

SMILES string

C[C@H](CC1)CO[C@]21O[C@@]3([H])C[C@@]4([H])[C@]5([H])CC=C6C[C@@H](O)C[C@@H](O[C@]7([H])O[C@H](C)[C@H](O)[C@H](O[C@]8([H])OC[C@@H](O)[C@H](O)[C@H]8O)[C@H]7O[C@]9([H])O[C@@H](C)[C@H](O)[C@@H](O)[C@H]9O)[C@]6(C)[C@@]5([H])CC[C@]4(C)[C@@]3([H])[C@@H]2C

InChI

1S/C44H70O16/c1-18-9-12-44(54-16-18)19(2)30-28(60-44)15-26-24-8-7-22-13-23(45)14-29(43(22,6)25(24)10-11-42(26,30)5)57-41-38(59-40-36(52)34(50)31(47)20(3)55-40)37(32(48)21(4)56-41)58-39-35(51)33(49)27(46)17-53-39/h7,18-21,23-41,45-52H,8-17H2,1-6H3/t18-,19+,20+,21-,23-,24-,25+,26+,27-,28+,29-,30+,31+,32+,33+,34-,35-,36-,37+,38-,39+,40+,41+,42+,43+,44-/m1/s1

InChI key

FHKHGNFKBPFJCB-LYLKFOBISA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Ophiopogonin D is a steroidal glycoside extracted from Ophiopogon japonicas that promotes antioxidative protection of the cardiovascular system. Ophiopogonin D protects the heart against doxorubicin-induced autophagic injury by reduction of both ROS generation and the disruption of the mitochondrial membrane damage.
Ophiopogonin D is a steroidal glycoside; antioxidant; protect cardiomyocytes from doxorubicin-induced toxicity.

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue Zhu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 69, 104973-104973 (2020-08-21)
Ophiopogonin D, a steroidal glycoside extracted from the Traditional Chinese Medicine Ophiopogon japonicus, shows anti-tumor property in several lines of cancers; however, its effect on triple-negative breast cancer (TNBC) has not been investigated. In this study, the anti-metastatic effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service