Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL80429

Dihydroberberine

phyproof® Reference Substance

Synonym(s):

5,8-Dihydro-9,10-dimethoxy-6H-benzo[g]-1,3-benzodioxolo[5,6-a]quinolizine, Dihydroumbellatine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19NO4
CAS Number:
Molecular Weight:
337.37
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

format

neat

InChI

1S/C20H19NO4/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2/h3-4,7-9H,5-6,10-11H2,1-2H3

InChI key

FZAGOOYMTPGPGF-UHFFFAOYSA-N

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • (+)-Abscisic acid in plant stress hormone research: Investigated the cloning of CAT genes in Satsuma mandarin and their expression characteristics in response to environmental stress and arbuscular mycorrhizal fungi. This research highlights the crucial role of (+)-abscisic acid in mediating plant responses to abiotic stresses, demonstrating its utility as a high-purity biochemical assay reagent for studying stress response signaling in plants (Liu et al., 2024).

  • (+)-Abscisic acid in biochemical assays: The physiological and biochemical response of Styrax tonkinensis seedlings to waterlogging stress was analyzed. This research underscores the importance of (+)-abscisic acid in biochemical assays aimed at understanding how plants cope with water stress, reinforcing its role in signal transduction studies related to plant survival mechanisms under adverse conditions (Chen et al., 2024).

  • Synthetic (+)-abscisic acid in plant biology: Examined RNA splicing and its effects on the postharvest physiological deterioration of cassava storage roots. The study discusses the involvement of (+)-abscisic acid in plant biology, particularly in how it influences postharvest changes through its role in signaling pathways that control physiological processes in plants (Gu et al., 2024).



Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service