Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

PHL80178

Amarogentin

phyproof® Reference Substance

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H30O13
CAS Number:
Molecular Weight:
586.54
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)OC(=O)c4c(cc(cc4c5cc(ccc5)O)O)O)O[C@@H]2OC=C3[C@H]([C@H]2C=C)CCOC3=O

InChI

1S/C29H30O13/c1-2-16-17-6-7-38-26(36)19(17)12-39-28(16)42-29-25(24(35)23(34)21(11-30)40-29)41-27(37)22-18(9-15(32)10-20(22)33)13-4-3-5-14(31)8-13/h2-5,8-10,12,16-17,21,23-25,28-35H,1,6-7,11H2/t16-,17+,21-,23-,24+,25-,28+,29+/m1/s1

InChI key

DBOVHQOUSDWAPQ-WTONXPSSSA-N

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xian-Ju Huang et al.
Frontiers in pharmacology, 11, 575772-575772 (2020-10-20)
Based on the pathological theory of lipid metabolism and using network pharmacology, this study was designed to investigate the protective effect of water extract of Veratrilla baillonii (WVBF) on non-alcoholic fatty liver disease (NAFLD) model using LO2 cells and to
Dejene Disasa et al.
Oxidative medicine and cellular longevity, 2020, 3184019-3184019 (2020-08-25)
In the present study, the replicative lifespan assay of yeast was used to guide the isolation of antiaging substance from Gentiana rigescens Franch, a traditional Chinese medicine. A compound with antiaging effect was isolated, and the chemical structure of this
Prabhjot Kaur et al.
BMC plant biology, 20(1), 225-225 (2020-05-21)
In this study, response surface methodology (RSM) and artificial neural network (ANN) was used to construct the predicted models of linear, quadratic and interactive effects of two independent variables viz. salicylic acid (SA) and chitosan (CS) for the production of
Gordana Joksic et al.
Current vascular pharmacology, 19(4), 359-369 (2020-05-30)
Gentiana lutea (GL), commonly known as yellow gentian, bitter root, and bitterwort, belongs to family Gentianaceae. GL belongs to genus Gentiana, which is a rich natural source of iridoids, secoiridoids, xantones, flavonoids, triterpenoids, and carbohydrates. Medicinal plants from Gentiana species

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service