Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NISTRM8446

Perfluorinated carboxylic acids and perfluorooctane sulfonamide in methanol

NIST® RM 8446

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

manufacturer/tradename

NIST®

format

multi-component solution

Other Notes

Reference/Informational value is provided for the following analytes.
Perfluorinated carboxylic acid (PFCAs) and perfluorooctane sulfonamide (PFOSA) compound: Perfluorohexanoic acid (PFHxA), Perfluoroheptanoic acid(PFHpA), Perfluorooctanoate acid(PFOA), Perfluorononanoic acid(PFNA), Perfluorodecanoic acid(PFDA), Perfluoroundecanoic acid(PFUnA), Perfluorododecanoic acid(PFDoA), Perfluorotridecanoic acid(PFTriA), Perfluorotetradecanoic acid (PFTA), Perfluorobutanoic acid(PFBA), Perfluoropentanoic acid(PFPeA) Perfluorooctane sulfonamide(PFOSA)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service