Skip to Content
MilliporeSigma
All Photos(1)

Documents

NISTRM8017

Polyvinylpyrrolidone coated silver nanoparticles

NIST® RM 8017, nominal diameter 75 nm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

manufacturer/tradename

NIST®

General description

One unit of RM 8017 comprises five serum vials, each consisting of lyophilized polyvinylpyrrolidone (PVP)-coated silver (Ag) nanoparticle cake. Every vial is vacuum sealed and contains approximately 2 mg of Ag and 20 mg of PVP (with a molar mass of approximately 40 kDa). For more information, please refer to the SDS and COA.
SRM 8017_cert

SRM 8017_SDS

Application

The Reference Material (RM) is intended for use as a benchmark and investigative tool to evaluate potential environmental, health, and safety risks associated with manufactured nanomaterials throughout their product life cycle.

Features and Benefits

  • The reference values for the mean silver particle size are traceable to the SI unit of length, expressed in nanometers, and are determined using the following methods:
    1) Atomic Force Microscopy (AFM)
    2) Transmission Electron Microscopy (TEM)
    3) Ultra-Small-Angle X-ray Scattering (USAXS)
    4) Dynamic Light Scattering (DLS)
  • The reference mass values for silver (Ag) are provided, and values are metrologically traceable to the SI unit of mass, expressed in milligrams.
  • Information values for copper, gold, silicon, and Iiron are provided and determined using ICP-MS method.
  • Expiration details, instructions for handling, storage and use are provided on the NIST certificate.

Other Notes

Reference/Informational value is provided for the following analytes.
Silver (Ag), Copper (Cu), Gold (Au), Silcon (Si), Iron (Fe)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service