Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NISTC2417

Lead-base alloy

NIST® SRM® C2417

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of disk

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is intended for use as a composition standard for optical emission spectrometric methods of analysis. It is in the form of a disk 50 mm (2 in.) in diameter and 16 mm ( 5/8 in. ) thickness. For more information, please refer to the COA and SDS.

SRM 2417_cert

SRM 2417_SDS

Other Notes

Certified for the analytes listed below.
Antimony (Sb), Arsenic (As), Bismuth (Bi), Copper (Cu), Silver (Ag)

Reference/Informational value is provided for the following analytes.
Aluminium (Al), Cadmium (Cd), Calcium (Ca), Cobalt (Co), Iron (Fe), Manganese (Mn), Nickel (Ni), Sulfur (S), Tellurium (Te), Tin (Sn), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 1A - STOT RE 1 Oral

Target Organs

Central nervous system,Blood,Immune system,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service