Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST916B

Bilirubin

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H36N4O6
CAS Number:
Molecular Weight:
584.66
Beilstein:
74376
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.24
Pricing and availability is not currently available.

packaging

pkg of 100 mg

manufacturer/tradename

NIST®

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

CC1=C(C=C)\C(NC1=O)=C\c2[nH]c(Cc3[nH]c(\C=C4/NC(=O)C(C=C)=C4C)c(C)c3CCC(O)=O)c(CCC(O)=O)c2C

InChI

1S/C33H36N4O6/c1-7-20-19(6)32(42)37-27(20)14-25-18(5)23(10-12-31(40)41)29(35-25)15-28-22(9-11-30(38)39)17(4)24(34-28)13-26-16(3)21(8-2)33(43)36-26/h7-8,13-14,34-35H,1-2,9-12,15H2,3-6H3,(H,36,43)(H,37,42)(H,38,39)(H,40,41)/b26-13-,27-14-

InChI key

BPYKTIZUTYGOLE-IFADSCNNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 916B consists of 100 mg of bilirubin.

SRM 916B_cert

SRM 916B_SDS

Application

Bilirubin standard reference material (SRM) is intended for use in the calibration and standardization of measurement procedures for the determination of bilirubin in clinical samples, it is also used for assigning bilirubin quantity values to in-house control materials.

Biochem/physiol Actions

Well over 99% of total bilirubin is transported as a conjugate with albumin. Abnormally high levels of bilirubin can cause severe neurological damage, but mildly elevated levels are linked to protection from oxidative stress.

Features and Benefits

  • SRM 916B is available with a certificate documenting NIST-certified mass fraction values the unconjugated Bilirubin.
  • Certified value is determined using a quantitative 1H-nuclear magnetic resonance spectroscopy (q1 H-NMR). 
  • Certified values are traceable to the International System of Units (SI) unit of mass, expressed as mass fraction percent. 
  • The NIST certificate is provided with safety, storage and use instructions.

Other Notes

Certified for the analytes listed below.
Bilirubin (C33H36N4O6)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service