Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NIST914B

Creatinine

NIST® SRM® 914b

Synonym(s):

2-Amino-1-methyl-2-imidazolin-4-one, 2-Imino-1-methylimidazolidin-4-one, 2-Imino-N-methylhydantoin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7N3O
CAS Number:
Molecular Weight:
113.12
Beilstein:
112061
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

packaging

pkg of 10 g

manufacturer/tradename

NIST®

technique(s)

NMR: suitable

mp

295 °C (dec.) (lit.)

application(s)

clinical testing

format

neat

SMILES string

CN1CC(=O)N=C1N

InChI

1S/C4H7N3O/c1-7-2-3(8)6-4(7)5/h2H2,1H3,(H2,5,6,8)

InChI key

DDRJAANPRJIHGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is certified as a chemical of known purity. It is intended primarily for use in the calibration and standardization of procedures used for determination of creatinine concentration and for routine evaluations of daily working standards used in these procedures. For more information, please refer to the SDS and COA.

SRM 914B_cert

SRM 914B_SDS

Other Notes

Certified for the analytes listed below.
Creatinine

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

554.0 °F - closed cup

Flash Point(C)

290 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service