Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST690

Iron ore, Canada

NIST® SRM® 690

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 100 g

manufacturer/tradename

NIST®

format

matrix material

General description

This Standard Reference Material (SRM) is in the form of a fine powder ( 0.1 mm) and is for use in checking chemical methods of analysis and in calibration with instrumental methods of analysis. For more information, please refer to the COA and SDS.

SRM 690_cert

SRM 690_SDS

Other Notes

Certified for the analytes listed below.
Aluminum Oxide (Al2O3), Calcium Oxide (CaO), Iron (Fe), Magnesium Oxide (MgO), Manganese(II) Oxide (MnO), Phosphorus (P), Potassium Oxide (K2O),
Sodium Oxide (Na2O), Silicon Dioxide (SiO2), Titanium Dioxide (TiO2)

Reference/Informational value is provided for the following analytes.
Chromium (Cr), Sulfur (S), Vanadium (V)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service