Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST3182

Chloride anion standard solution

NIST® SRM® 3182

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

This Standard Reference Material (SRM) is intended as a primary standard for the quantitative determination of chloride using anion ion chromatography (IC) or other methods. A unit of SRM 3182 consists of five 10 mL sealed borosilicate glass ampoules of solution. The solution is prepared gravimetrically to contain a known mass fraction of chloride dissolved in filtered (0.22 μm) water having a minimum resistivity of 18 MΩcm. For more information, plese refer to the SDS and COA.

SRM 3182_cert

SRM 3182_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Chloride (Cl-)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service