Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

NIST2892

Ethanol-water solution

NIST® SRM® 2892, nominal mass fraction 0.04%

Synonym(s):

Ethanol solution, Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 1.2 mL

manufacturer/tradename

NIST®

technique(s)

Karl-Fischer titration: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is a solution of ethanol in water at a nominal mass fraction of 0.04 % and is intended primarily for use in the calibration of instruments and techniques used for the determination of ethanol. A unit of SRM 2892 consists of five 2 mL ampoules, each containing approximately 1.2 mL of solution.For more information, please refer to the SDS and COA.

SRM 2892_cert

SRM 2892_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Ethanol (C2H6O)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service