Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2787

Fine particulate matter

NIST® SRM® 2787, <10 μm

Synonym(s):

Dust

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 1 bottle

manufacturer/tradename

NIST®

application(s)

industrial hygiene

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

Each unit of SRM 2787 contains a bottle with a quantity of particulate matter ranging from 100 mg to 140 mg. For more information, please refer to the SDS and COA.
SRM 2787_cert
SRM 2787_SDS

Application

Fine particulate matter Standard Reference Material (SRM) is intended primarily for the evaluation of analytical methods used in the determination of selected polycyclic aromatic hydrocarbons (PAHs), nitro-substituted PAHs (nitro-PAHs), polybrominated diphenyl ether (PBDE) congeners, hexabromocyclododecane (HBCD) isomers, sugars, polychlorinated dibenzo-p-dioxin (PCDD) and dibenzofuran (PCDF) congeners, inorganic constituents, and particle-size characteristics in atmospheric particulate material and similar matrices.

Features and Benefits

  • The certified mass fraction values for PAHs, PBDE congeners, and specific inorganic components are reported and are metrologically traceable to SI units of mass.
  • Accomplished with non-certified mass fraction values for nitro-PAHs, PBDE congeners, sugars, HBCD isomers, and particle-size characteristics.
  • Period of Validity, instructions for handling, storage, and use are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Fluorene (C13H10), Fluoranthene (C16H10), Pyrene (C16H10), Retene (C18H18), Benzo[ghi]fluoranthene (C18H10), Chrysene (C18H12), Triphenylene (C6H4)3), Benzo[b]fluoranthene (C20H12), Benzo[j]fluoranthene (C20H12), Benzo[k]fluoranthene (C20H12), Benzo[a]fluoranthene (C20H12), Benzo[e]pyrene (C20H12), Benzo[a]pyrene (C20H12), Perylene (C20H12), Benzo[ghi]perylene (C22H12), Indeno[1,2,3-cd]pyrene (C22H12), Dibenz[a,c]anthracene (C22H14), Dibenz[a,j]anthracene (C22H14), Benzo[b]chrysene (C22H14), Picene (C22H14), Coronene (C24H12), Dibenzo[b,k]fluoranthene (C24H14), Dibenzo[a,e]pyrene (C24H14), PBDE 99 (2,2′,4,4′,5-Pentabromodiphenyl ether), PBDE 209 (Decabromodiphenyl ether), Mercury (Hg), Aluminum (Al), Cadmium (Cd), Chromium (Cr), Lead (Pb), Manganese (Mn), Vanadium (V)

Reference/Informational value is provided for the following analytes.
1-Nitropyrene, 2-Nitrofluoranthene, 9-Nitroanthracene, 3-Nitrophenanthrene, 9-Nitrophenanthrene, 3-Nitrofluoranthene, 7-Nitrobenz[a]anthracene, Naphthalene, 1-Methylnaphthalene, 2-Methylnaphthalene, 1,6-Dimethylnaphthalene, Acenaphthylene, 1-Methylphenanthrene, 2-Methylanthracene, 4H-cyclopenta[def]phenanthrene, 1-Methylfluoranthene, 3-Methylfluoranthene, 4-Methylpyrene, Cyclopenta[cd]pyrene, 3-Methylchrysene, 6-Methylchrysene, Dibenz[a,h]anthracene, Dibenzo[a,h]pyrene
PFE : Biphenyl, Dibenzothiophene, Phenanthrene, Anthracene, 2-Methylphenanthrene, 3-Methylphenanthrene, 9-Methylphenanthrene, 1,7-Dimethylphenanthrene, 1-Methylpyrene, Benzo[c]phenanthrene, Benz[a]anthracene, Biphenyl, Dibenzothiophene, Phenanthrene, Anthracene, 2-Methylphenanthrene, 3-Methylphenanthrene, 9-Methylphenanthrene, 1,7-Dimethylphenanthrene, 1-Methylpyrene, Benzo[c]phenanthrene, Benz[a]anthracene, PBDE 47 (2,2′,4,4′-Tetrabromodiphenyl ether), PBDE 100 (2,2′,4,4′,6-Pentabromodiphenyl ether), PBDE 153 (2,2′,4,4′,5,5′-Hexabromodiphenyl ether), PBDE 154 (2,2′,4,4′,5,6′-Hexabromodiphenyl ether), PBDE 191 (2,3,3′,4,4′,5′,6-Heptabromodiphenyl ether, Antimony (Sb), Arsenic (As), Calcium (Ca), Cesium (Ce), Chlorine (Cl), Copper (Cu), Cobalt (Co), Iron (Fe), Lanthanum (La), Nickel (Ni), Samarium (Sm), Sodium (Na), Titanium (Ti), Zinc (Zn), Levoglucosan,Mannosan, Glactosan

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service