Skip to Content
MilliporeSigma
All Photos(1)

Documents

NIST2690

Coal fly ash

NIST® SRM® 2690

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 3 x 10 g

manufacturer/tradename

NIST®

application(s)

environmental

format

matrix material

Looking for similar products? Visit Product Comparison Guide

General description

This Standard Reference Material (SRM) is intended for use in the evaluation of analytical methods used for the classification of coal fly ash and for the determination of constituent elements in coal fly ash or materials of a similar matrix. A unit of SRM 2690 consists of three 10 g hermetically sealed glass vials of fly ash pulverized to less than 150 μm particle size and blended to a high degree of homogeneity. For more information, please refer to the SDS and COA.

SRM 2690_cert
SRM 2690_SDS

Other Notes

Certified for the analytes listed below.
Aluminum (Al), Calcium (Ca), Iron (Fe), Magnesium (Mg), Phosphorus (P), Potassium (K), Silicon (Si), Sodium (Na), Sulfur (S), Titanium (Ti)

Reference/Informational value is provided for the following analytes.

Antimony (Sb), Arsenic (As), Barium (Ba), Beryllium (Be), Cobalt (Co), Chromium (Cr), Cesium (Cs), Europium (Eu), Hafnium (Hf), Lead (Pb), Manganese (Mn), Mercury (Hg), Nickel (Ni), Scandium (Sc), Selenium (Se), Strontium (Sr), Thorium (Th), Zinc (Zn)

See certificate for values and more details at nist.gov/SRM

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service