Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

NIST2031C

Metal-on-Fused-Silica Neutral Density Filters (250 nm to 635 nm)

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
SiO2
CAS Number:
Molecular Weight:
60.08
UNSPSC Code:
41116107
NACRES:
NA.24

packaging

pkg of 100 g

manufacturer/tradename

NIST®

application(s)

general analytical

storage temp.

15-25°C

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

This Standard Reference Material (SRM) is intended for use in the verification of the transmittance and absorbance scales of spectrophotometers in the ultraviolet and visible spectral regions. SRM 2031c is a transfer standard certified using the NIST Materials Measurement Laboratory Transfer Spectrophotometer (MMLTS) with traceability through the NIST second generation High Accuracy Spectrophotometer (HAS II), modeled after the original instrument [2]. A unit consists of three individual neutral-density filters in separate metal holders and one empty filter holder, all stored in a black anodized-aluminum container.

SRM 2031C_cert

SRM 2031C_SDS

Other Notes

Please download a current certificate at nist.gov/SRM for current analytes and certified values.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service