Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

N1511

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide hydrate

≥99%

Synonym(s):

β-DPN, β-NAD, Coenzyme 1, Cozymase, DPN, Diphosphopyridine nucleotide, NAD, Nadide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7O14P2 · xH2O
CAS Number:
Molecular Weight:
663.43 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
39200202
PubChem Substance ID:
NACRES:
NA.55

biological source

Saccharomyces cerevisiae

Quality Level

Assay

≥99%

form

crystalline powder

color

white to off-white

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2.O[C@@H]3[C@@H](COP(O)(OP(OC[C@@H](O4)[C@@H](O)[C@@H](O)[C@H]4[N+]5=CC=CC(C(N)=O)=C5)([O-])=O)=O)OC[C@@H]3O

InChI

1S/C21H27N7O14P2/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33/h1-4,7-8,10-11,13-16,20-21,29-32H,5-6H2,(H5-,22,23,24,25,33,34,35,36,37)/t10-,11-,13-,14-,15-,16-,20-,21-/m1/s1

InChI key

BAWFJGJZGIEFAR-NNYOXOHSSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide hydrate has been used:
  • to prepare NAD+ standard solution for a study on NAD+ metabolomics
  • as a component of MurI assay reaction partial mixture for glutamate racemase reaction
  • as a component of a chromogenic staining solution to stain zebrafish embryos

Biochem/physiol Actions

β-Nicotinamide adenine dinucleotide (β-NAD) is a cofactor of alcohol dehydrogenase and acts as an inhibitory neurotransmitter in visceral smooth muscles. The NAD/NADH ratio has a role in the regulation of intracellular redox potential. It thereby influences metabolic reactions in vivo.
Electron acceptor

Packaging

Packaged by solid weight.

Other Notes

Crystalline NAD contains less than 0.1% (w/w) residual organic solvent, is less hygroscopic and electrostatic, and is more stable at ambient temperature than amorphous NAD.
This is the common form of NAD.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

beta-Nicotinamide adenine dinucleotide is an inhibitory neurotransmitter in visceral smooth muscle
Mutafova-Yambolieva VN, et al.
Proceedings of the National Academy of Sciences of the USA (2007)
Assays for NAD+-Dependent Reactions and NAD+ Metabolites
Schultz MB, et al.
Methods in Molecular Biology, 77-90 (2018)
Determination of the cytosolic NADPH/NADP ratio in Saccharomyces cerevisiae using shikimate dehydrogenase as sensor reaction
Zhang J, et al.
Scientific Reports (2015)
The zebrafish kidney mutant zeppelin reveals that brca2/fancd1 is essential for pronephros development
Kroeger Jr PT, et al.
Developmental Biology (2017)
Detection of D-glutamate Production from the Dual Function Enzyme, 4-amino-4-deoxychorismate Lyase/D-amino Acid Transaminase, in Mycobacterium smegmatis
Opel-Reading HK, et al.
Bio-protocol (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service