Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N10

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 14.42 mPa.s (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

14.42 mPa.s(25 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon General Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F

Flash Point(C)

23 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

AmirAbbas YousefiAmin et al.
ACS nano, 13(2), 2389-2397 (2019-02-02)
Colloidal nanocrystals from PbS are successfully applied in highly sensitive infrared photodetectors with various device architectures. Here, we demonstrate all-printed devices with high detectivity (∼1012 cm Hz1/2/W) and a cut-off frequency of >3 kHz. The low material consumption (<0.3 mg
Goutham R Adelli et al.
Drug delivery, 24(1), 370-379 (2017-02-07)
The goal of the present study is to develop polymeric matrix films loaded with a combination of free diclofenac sodium (DFS Effect of ratio of DFS and IR on the DFS:IR complexation efficiency was studied using batch processing. DFS:IR complex
Yanfeng Liu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 65, 104778-104778 (2020-01-22)
The need of in vitro alternative methods has been increasing in toxicology research as well as in cosmetic industry in China recently. Following the establishment of China EpiSkin™ skin corrosion and irritation testing methods, both as stand-alone in vitro tests

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service