Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M187

Supelco

(+)-3-Methoxymorphinan hydrochloride

analytical standard, for drug analysis

Synonym(s):

(+)-cis-1,3,4,9,10,10a-Hexahydro-6-methoxy-2H-10,4a-iminoethanophenanthrene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO · HCl
CAS Number:
Molecular Weight:
293.83
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

form

powder

optical activity

[α]/D +21.0 to +24.0°, c = 1 in methanol

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white

solubility

H2O: 10 mg/mL, clear

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

[H][C@]12[C@@]3(CCCC2)C4=C(C=CC(OC)=C4)C[C@]1([H])NCC3.Cl

InChI

1S/C17H23NO.ClH/c1-19-13-6-5-12-10-16-14-4-2-3-7-17(14,8-9-18-16)15(12)11-13;/h5-6,11,14,16,18H,2-4,7-10H2,1H3;1H/t14-,16+,17+;/m1./s1

InChI key

IBNAJETZECTKHN-YPYJQMNVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Franck Merlier et al.
The Analyst, 142(19), 3747-3757 (2017-09-12)
A microfluidic liver biochip was coupled with a mass spectrometer to detect in real time the drug metabolism of hepatocytes. The hepatocytes were cultivated in the biochip for 35 h. The biochip was placed in a small-scale incubator in which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service