Skip to Content
MilliporeSigma
All Photos(1)

Documents

M1825010

Metoclopramide impurity E

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N,N-Diethylethylenediamine, 2-Diethylaminoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCH2CH2NH2
CAS Number:
Molecular Weight:
116.20
Beilstein:
605299
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metoclopramide

manufacturer/tradename

EDQM

refractive index

n20/D 1.436 (lit.)

bp

145-147 °C (lit.)

density

0.827 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCN(CC)CCN

InChI

1S/C6H16N2/c1-3-8(4-2)6-5-7/h3-7H2,1-2H3

InChI key

UDGSVBYJWHOHNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metoclopramide impurity E EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

89.6 °F - closed cup

Flash Point(C)

32 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng-Yun J Huang et al.
International journal of nanomedicine, 10, 463-473 (2015-01-28)
In this study, the (188)Re-labeled PEGylated nanoliposome ((188)Re-liposome) was prepared and evaluated as a therapeutic agent for glioma. The reporter cell line, F98(luc) was prepared via Lentivector expression kit system and used to set up the orthotopic glioma-bearing rat model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service