Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M1808000

Metixene hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methixene monohydrochloride monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NSl · HCl · H2O
CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

metixene

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

S1c2c(cccc2)C(c4c1cccc4)CC3CN(CCC3)C.[Cl-].O.[H+]

InChI

1S/C20H23NS.ClH.H2O/c1-21-12-6-7-15(14-21)13-18-16-8-2-4-10-19(16)22-20-11-5-3-9-17(18)20;;/h2-5,8-11,15,18H,6-7,12-14H2,1H3;1H;1H2

InChI key

RAOHHYUBMJLHNC-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Metixene hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

JAMA, 195(10), 851-851 (1966-03-07)
Methixene hydrochloride, an anticholinergic agent, may be useful in the symptomatic treatment of functional bowel disorders. Adverse effects reported from its use are those of the anticholinergic drugs in general.
[An anticholinergic agent, the side effect of mouth dryness and hazards for the teeth].
W H van Palenstein Helderman et al.
Nederlands tijdschrift voor geneeskunde, 127(51), 2318-2321 (1983-12-17)
M I Walash et al.
Pharmaceutisch weekblad. Scientific edition, 8(4), 234-238 (1986-08-22)
A simple and rapid colorimetric method for the quantitative determination of the thioxanthene derivatives, chlorprothixene, tiotixene and metixene has been performed either in their pure form or in tablets. The procedure is based upon the formation of charge transfer complexes
P A Fischer et al.
Journal of neural transmission. Supplementum, 25, 137-147 (1987-01-01)
To quantify the clinical benefit of an additional deprenyl administration in L-dopa pretreated patients we have performed a study of 30 patients suffering from advanced parkinsonism. During the first phase of the study over three months under controlled conditions deprenyl
[Treatment of extrapyramidal tremors in the aged].
D Hust
Die Medizinische Welt, 32(19), 743-746 (1981-05-08)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service