Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

L4408

Sigma-Aldrich

Lithium chloride

BioXtra, ≥99.0% (titration)

Synonym(s):

LiCl, Lithium Chloride Solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder
solubility:
H2O: 1 M, clear, colorless

grade

for analytical purposes

Quality Level

product line

BioXtra

Assay

≥99.0% (titration)

form

powder

impurities

≤0.002% Phosphorus (P)
≤0.1% Insoluble matter

mp

605 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

sulfate (SO42-): ≤0.01%

cation traces

Al: ≤0.0005%
Ba: ≤0.002%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.002%
K: ≤0.005%
Mg: ≤0.005%
NH4+: ≤0.1%
Na: ≤0.005%
Pb: ≤0.001%
Zn: ≤0.001%

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • WDR1 promotes prostate cancer progression through Wnt/β-catenin signaling.: Although this study primarily focuses on WDR1′s role in cancer, the use of Lithium Chloride in Wnt signaling pathway modulation offers insights into its potential therapeutic applications in cancer treatment (Cheng J et al., 2024) (Cheng J et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa M Leon et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 44(10), 1742-1751 (2019-04-09)
The overlap in neurobiological circuitry mediating the physiological and behavioral response to satiation and noxious/stressful stimuli are not well understood. The interaction between serotonin (5-HT) and glucagon-like peptide-1 (GLP-1) could play a role as upstream effectors involved in mediating associations
Jong Sung Kim et al.
Nature communications, 10(1), 3575-3575 (2019-08-10)
The direct sensing and storing of the information of liquids with different polarities are of significant interest, in particular, through means related to human senses for emerging biomedical applications. Here, we present an interactive platform capable of sensing and storing
Marie Goutierre et al.
Cell reports, 28(1), 91-103 (2019-07-04)
KCC2 regulates neuronal transmembrane chloride gradients and thereby controls GABA signaling in the brain. KCC2 downregulation is observed in numerous neurological and psychiatric disorders. Paradoxical, excitatory GABA signaling is usually assumed to contribute to abnormal network activity underlying the pathology.
Alessandro Di Maio et al.
EvoDevo, 6, 17-17 (2015-07-15)
Wnt signaling is one of the earliest and most highly conserved regulatory pathways for the establishment of the body axes during regeneration and early development. In regeneration, body axes determination occurs independently of tissue rearrangement and early developmental cues. Modulation
Karla M Acevedo et al.
The Journal of biological chemistry, 289(16), 11007-11019 (2014-03-13)
Amyloid precursor protein (APP) undergoes post-translational modification, including O- and N-glycosylation, ubiquitination, and phosphorylation as it traffics through the secretory pathway. We have previously reported that copper promotes a change in the cellular localization of APP. We now report that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service